Assay Details
| Gene Symbol: MRPL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014763.3 | NP_055578.2 | 2 - 3 | 251 | 72 | |
| GenBank mRNA | AK291250.1 | - | 2 - 3 | 237 | 72 | |
| AK295578.1 | - | 2 - 3 | 223 | 72 | ||
| BC030144.2 | - | 2 - 3 | 236 | 4763405 | 72 | |
| BI551514.1 | - | 2 - 3 | 256 | 72 | ||
| BQ432684.1 | - | 2 - 3 | 254 | 72 | ||
| D14660.1 | - | 2 - 3 | 224 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000476.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK295578.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000604845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007411.hg.1 | ENST00000393909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007411.hg.1 | ENST00000409374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007411.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007411.hg.1 | TC02000476.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007408.hg.1 | ENST00000393909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007408.hg.1 | ENST00000409374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007408.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007408.hg.1 | TC02000476.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007413.hg.1 | ENST00000393909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007413.hg.1 | ENST00000409374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007413.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007413.hg.1 | TC02000476.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156428.hg.1 | TC0200008128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156430.hg.1 | TC0200008128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156432.hg.1 | TC0200008128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000604845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |

MRPL19
Genomic Map