Assay Details
| Gene Symbol: GBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000158.3 | NP_000149.3 | 2 - 3 | 597 | 95 | |
| GenBank mRNA | AB209731.1 | - | 2 - 3 | 487 | 95 | |
| AK125918.1 | - | 2 - 3 | 960 | 95 | ||
| AK300620.1 | - | 2 - 3 | 244 | 95 | ||
| BC012098.1 | - | 2 - 3 | 526 | 4574938 | 95 | |
| L07956.1 | - | 2 - 3 | 404 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001575.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028572.hg.1 | ENST00000264326.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028572.hg.1 | ENST00000429644.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028572.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028572.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028572.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028572.hg.1 | TC03001575.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186892.hg.1 | TC0300011651.hg.1 | Clariom D TC |

GBE1
Genomic Map