Assay Details
| Gene Symbol: GBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000158.3 | NP_000149.3 | 5 - 6 | 975 | 131 | |
| GenBank mRNA | AB209731.1 | - | 5 - 6 | 865 | 131 | |
| AK125918.1 | - | 5 - 6 | 1338 | 131 | ||
| AK297939.1 | - | 3 - 4 | 408 | 131 | ||
| AK300620.1 | - | 5 - 6 | 622 | 131 | ||
| AK312245.1 | - | 5 - 6 | 824 | 131 | ||
| BC012098.1 | - | 5 - 6 | 904 | 4574938 | 131 | |
| L07956.1 | - | 5 - 6 | 782 | 131 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001575.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297939.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028562.hg.1 | ENST00000264326.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028562.hg.1 | ENST00000429644.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028562.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028562.hg.1 | ENST00000536832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028562.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028562.hg.1 | TC03001575.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028563.hg.1 | ENST00000264326.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028563.hg.1 | ENST00000429644.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028563.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028563.hg.1 | ENST00000536832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028563.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028563.hg.1 | TC03001575.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186884.hg.1 | TC0300011651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000477426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000484687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000486920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011651.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | ENST00000429644.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | ENST00000489715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | ENST00000498468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | NM_000158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186883.hg.1 | TC0300011651.hg.1 | Clariom D TC |

GBE1
Genomic Map