Assay Details
| Gene Symbol: STAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324282.1 | NP_001311211.1 | 2 - 3 | 338 | 99 | |
| NM_001324288.1 | NP_001311217.1 | 2 - 3 | 338 | 99 | ||
| NM_003473.3 | NP_003464.1 | 2 - 3 | 338 | 99 | ||
| XM_011519695.2 | XP_011517997.1 | 2 - 3 | 314 | 99 | ||
| GenBank mRNA | AK314405.1 | - | 2 - 3 | 222 | 99 | |
| AU118708.1 | - | 2 - 3 | 339 | 99 | ||
| BC030586.2 | - | 2 - 3 | 281 | 4820857 | 99 | |
| U43899.1 | - | 2 - 3 | 165 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000129.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001324288.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
autophagy
signal transduction
positive regulation of signal transduction
endosomal transport
multivesicular body assembly
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of exosomal secretion
regulation of extracellular exosome assembly
SH3/SH2 adaptor activity
protein binding
intracellular protein transport
autophagy
signal transduction
positive regulation of signal transduction
endosomal transport
multivesicular body assembly
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of exosomal secretion
regulation of extracellular exosome assembly
SH3/SH2 adaptor activity
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10001413.hg.1 | ENST00000377524.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001413.hg.1 | ENST00000460949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001413.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001413.hg.1 | TC10000129.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000486183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000486183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000486183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000494250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000494250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000494250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | ENST00000494250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147236.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147236.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147236.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147236.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147236.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147236.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147236.hg.1 | TC1000006904.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000377524.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000445846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000486183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000486183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000486183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000494250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000494250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000494250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | ENST00000494250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006904.hg.1 | NM_003473.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 202172 | - | - |

STAM
Genomic Map