| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10001415.hg.1 |
ENST00000377524.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10001415.hg.1 |
ENST00000460949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10001415.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10001415.hg.1 |
TC10000129.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000486183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000486183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000486183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000494250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000494250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000494250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
ENST00000494250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
ENST00000486183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
ENST00000486183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000147238.hg.1 |
TC1000006904.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000377524.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000445846.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000486183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000486183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000486183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000494250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000494250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000494250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
ENST00000494250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006904.hg.1 |
NM_003473.3 |
Refseq |