| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011630.hg.1 |
ENST00000240874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011630.hg.1 |
ENST00000360013.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011630.hg.1 |
ENST00000460856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011630.hg.1 |
NM_001024660.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011630.hg.1 |
NM_003947.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011630.hg.1 |
TC03000642.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
ENST00000240874.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
ENST00000354186.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
ENST00000360013.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
ENST00000460856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
NM_001024660.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
NM_003947.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
ENST00000240874.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
ENST00000354186.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
ENST00000360013.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
ENST00000460856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
NM_001024660.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
NM_003947.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161233.hg.1 |
TC0300008594.hg.1 |
Clariom D TC |