Assay Details
| Gene Symbol: AP1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283.3 | NP_001274.1 | 3 - 4 | 404 | 85 | |
| GenBank mRNA | AB015319.1 | - | 3 - 4 | 321 | 85 | |
| AK312151.1 | - | 3 - 4 | 370 | 85 | ||
| BC003561.1 | - | 3 - 4 | 323 | 3504050 | 85 | |
| BI597121.1 | - | 3 - 4 | 409 | 85 | ||
| BI598615.1 | - | 3 - 4 | 409 | 85 | ||
| BT006779.1 | - | 2 - 3 | 294 | 85 | ||
More Information
Set Membership: |
|

AP1S1
Genomic Map