Assay Details
| Gene Symbol: NEK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304384.1 | NP_001291313.1 | 10 - 11 | 1223 | 77 | |
| NM_152534.4 | NP_689747.3 | 32 - 33 | 3323 | 77 | ||
| XM_006712998.2 | XP_006713061.1 | 32 - 33 | 3284 | 77 | ||
| XM_006712999.3 | XP_006713062.1 | 32 - 33 | 3284 | 77 | ||
| XM_017005761.1 | XP_016861250.1 | 31 - 32 | 3236 | 77 | ||
| XM_017005762.1 | XP_016861251.1 | 33 - 34 | 3400 | 77 | ||
| XM_017005763.1 | XP_016861252.1 | 32 - 33 | 3381 | 77 | ||
| XM_017005764.1 | XP_016861253.1 | 31 - 32 | 3213 | 77 | ||
| XM_017005765.1 | XP_016861254.1 | 31 - 32 | 3185 | 77 | ||
| XM_017005767.1 | XP_016861256.1 | 30 - 31 | 3149 | 77 | ||
| XM_017005768.1 | XP_016861257.1 | 31 - 32 | 3146 | 77 | ||
| GenBank mRNA | AK057247.1 | - | 10 - 11 | 1175 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

NEK10
Genomic Map