Assay Details
| Gene Symbol: CD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153826.3 | NP_722548.1 | 11 - 12 | 1257 | 99 | |
| NM_172353.2 | NP_758863.1 | 10 - 11 | 1212 | 99 | ||
| NM_172359.2 | NP_758869.1 | 12 - 13 | 1302 | 99 | ||
| NM_172361.2 | NP_758871.1 | 9 - 10 | 1170 | 99 | ||
| XM_011509563.1 | XP_011507865.1 | 11 - 12 | 1260 | 99 | ||
| XM_011509564.1 | XP_011507866.1 | 10 - 11 | 1215 | 99 | ||
| GenBank mRNA | AK129560.1 | - | 4 - 5 | 763 | 99 | |
| AK222822.1 | - | 12 - 13 | 1228 | 99 | ||
| AK291227.1 | - | 11 - 12 | 1211 | 99 | ||
| BC030594.2 | - | 11 - 12 | 1197 | 4823040 | 99 | |
| BX537451.1 | - | 8 - 9 | 5830 | 99 | ||
| BX640613.1 | - | 11 - 12 | 1239 | 99 | ||
| EF076055.1 | - | 11 - 12 | 1115 | 99 | ||
| EF076056.1 | - | 10 - 11 | 1070 | 99 | ||
| EF076057.1 | - | 9 - 10 | 1028 | 99 | ||
| EF076058.1 | - | 9 - 10 | 1028 | 99 | ||
| S51940.1 | - | 10 - 11 | 1124 | 99 | ||
| X59407.1 | - | 10 - 11 | 1141 | 99 | ||
| X59408.1 | - | 11 - 12 | 1227 | 99 | ||
| X59409.1 | - | 12 - 13 | 1231 | 99 | ||
| Y00651.1 | - | 11 - 12 | 1125 | 99 | ||
More Information
Set Membership: |
|

CD46
Genomic Map