Assay Details
| Gene Symbol: HMGCLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042406.1 | NP_001035865.1 | 4 - 5 | 558 | 99 | |
| NM_001287746.1 | NP_001274675.1 | 5 - 6 | 654 | 99 | ||
| NM_019036.2 | NP_061909.2 | 5 - 6 | 648 | 99 | ||
| XM_011514700.2 | XP_011513002.1 | 4 - 5 | 558 | 99 | ||
| GenBank mRNA | AK055075.1 | - | 5 - 6 | 539 | 99 | |
| AK127587.1 | - | 5 - 6 | 621 | 99 | ||
| BC024194.2 | - | 4 - 5 | 439 | 4818781 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

HMGCLL1
Genomic Map