Assay Details
| Gene Symbol: GPR149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038705.1 | NP_001033794.1 | 2 | 1218 | 72 | |
| GenBank mRNA | AY255534.1 | - | 2 | 653 | 72 | |
| DQ318360.1 | - | 2 | 1218 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GPR149
Genomic Map