Assay Details
| Gene Symbol: JAZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175061.3 | NP_778231.2 | 3 - 4 | 623 | 101 | |
| XM_006715656.1 | XP_006715719.1 | 4 - 5 | 499 | 101 | ||
| XM_017011831.1 | XP_016867320.1 | 3 - 4 | 391 | 101 | ||
| XM_017011832.1 | XP_016867321.1 | 3 - 4 | 323 | 101 | ||
| GenBank mRNA | AK091311.1 | - | 3 - 4 | 548 | 101 | |
| AL834234.1 | - | 2 - 3 | 342 | 101 | ||
| AY372319.1 | - | 3 - 4 | 401 | 101 | ||
| BC042441.1 | - | 3 - 5 | 411 | 4831063 | 101 | |
| BC047229.1 | - | 3 - 4 | 410 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001232.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07019255.hg.1 | ENST00000283928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019255.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019255.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07019255.hg.1 | TC07001232.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000420835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000420835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000420835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000420835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000420835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000420835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000420835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000420835.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000454041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010585.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000283928.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000430432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000427814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000447620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000452993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | ENST00000466516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | NM_175061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700175306.hg.1 | TC0700010585.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 210801 | - | - |

JAZF1
Genomic Map