Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 102 member A | FAM102A |
C9orf132, EEIG1, SYM-3A, bA203J24.7 |
Sense | Chr. 9 - 127940582 - 127980533 on NCBI Build GRCh38 | |
| Downstream | nuclear apoptosis inducing factor 1 | NAIF1 |
C9orf90, bA379C10.2 |
94.8kb | Sense | Chr. 9 - 128061233 - 128067321 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map