Assay Details
| Gene Symbol: SLC5A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178498.3 | NP_848593.2 | 7 - 8 | 1261 | 74 | |
| XM_006718155.3 | XP_006718218.1 | 7 - 8 | 1164 | 74 | ||
| XM_006718156.3 | XP_006718219.1 | 7 - 8 | 1500 | 74 | ||
| XM_011519920.2 | XP_011518222.1 | 8 - 9 | 741 | 74 | ||
| XM_011519921.2 | XP_011518223.1 | 7 - 8 | 1501 | 74 | ||
| XM_017017244.1 | XP_016872733.1 | 8 - 9 | 1187 | 74 | ||
| GenBank mRNA | AY299482.1 | - | 7 - 8 | 970 | 74 | |
| BC041454.1 | - | 7 - 8 | 730 | 74 | ||
| BC049207.1 | - | 7 - 8 | 1233 | 5188979 | 74 | |
| CN370367.1 | - | 3 - 4 | 370 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001506.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018844.hg.1 | ENST00000280467.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018844.hg.1 | ENST00000396005.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018844.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018844.hg.1 | TC11001506.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178768.hg.1 | TC1100010393.hg.1 | Clariom D TC |

SLC5A12
Genomic Map