Assay Details
| Gene Symbol: VSIG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC001437.1 | - | 1 - 2 | 297 | 138 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |

VSIG10
Genomic Map