| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18003967.hg.1 |
ENST00000327283.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18003967.hg.1 |
ENST00000353319.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18003967.hg.1 |
NM_001207013.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18003967.hg.1 |
NM_080422.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18003967.hg.1 |
NM_080423.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18003967.hg.1 |
TC18000377.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000327283.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000353319.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000585666.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000587703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000589444.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000591115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000591901.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
NM_001207013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
NM_080422.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
NM_080423.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
TC1800008128.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000327283.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000353319.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000585666.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000587703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000589444.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000591115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
ENST00000591901.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
NM_001207013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
NM_080422.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155631.hg.1 |
NM_080423.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000327283.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000353319.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000585666.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000589444.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000591115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000591901.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
NM_001207013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
NM_080422.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
NM_080423.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
TC1800008128.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000327283.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000353319.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000585666.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000589444.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000591115.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
ENST00000591901.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
NM_001207013.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
NM_080422.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155630.hg.1 |
NM_080423.2 |
Refseq |