Assay Details
| Gene Symbol: SIN3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145357.1 | NP_001138829.1 | 21 | 129 | ||
| NM_001145358.1 | NP_001138830.1 | 21 | 129 | |||
| NM_015477.2 | NP_056292.1 | 21 | 129 | |||
| XM_006720465.3 | XP_006720528.1 | 21 | 129 | |||
| XM_006720466.3 | XP_006720529.1 | 22 | 129 | |||
| XM_006720467.3 | XP_006720530.1 | 23 | 129 | |||
| GenBank mRNA | BM676304.1 | - | 1 | 166 | 129 | |
More Information
Set Membership: |
|

SIN3A
Genomic Map