| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000252229.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000399150.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000494577.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000494577.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000494577.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000494577.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000494577.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
ENST00000538442.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007585.hg.1 |
NM_001289161.1 |
Refseq |