Assay Details
| Gene Symbol: METTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC000550.1 | - | 6 | 1327 | 3163932 | 134 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000547653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000548681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000549773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000551117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | ENST00000553125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012815.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | ENST00000257848.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | NM_005371.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | NM_023033.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200203953.hg.1 | TC1200012815.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200203951.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203951.hg.1 | ENST00000324871.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200203951.hg.1 | TC1200012815.hg.1 | Clariom D TC |

METTL1
Genomic Map