Assay Details
| Gene Symbol: GH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000515.4 | NP_000506.2 | 2 - 3 | 247 | 133 | |
| GenBank mRNA | A00469.1 | - | 2 - 3 | 210 | 133 | |
| AF110644.1 | - | 2 - 3 | 181 | 133 | ||
| AF185611.1 | - | 2 - 3 | 189 | 133 | ||
| AV749353.1 | - | 2 - 3 | 247 | 133 | ||
| AY613431.1 | - | 2 - 3 | 233 | 133 | ||
| AY613432.1 | - | 2 - 3 | 233 | 133 | ||
| BC075012.2 | - | 2 - 3 | 198 | 30915404 | 133 | |
| BC075013.2 | - | 2 - 3 | 198 | 30915617 | 133 | |
| BC090045.1 | - | 2 - 3 | 214 | 30394115 | 133 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17002854.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor internalization
JAK-STAT cascade
positive regulation of activation of JAK2 kinase activity
positive regulation of phosphatidylinositol 3-kinase signaling
glucose transport
response to estradiol
positive regulation of multicellular organism growth
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
positive regulation of MAP kinase activity
positive regulation of insulin-like growth factor receptor signaling pathway
positive regulation of JAK-STAT cascade
positive regulation of peptidyl-tyrosine phosphorylation
growth hormone receptor signaling pathway
JAK-STAT cascade involved in growth hormone signaling pathway
bone maturation
growth hormone receptor binding
prolactin receptor binding
hormone activity
protein binding
growth factor activity
metal ion binding
positive regulation of receptor internalization
JAK-STAT cascade
positive regulation of activation of JAK2 kinase activity
positive regulation of phosphatidylinositol 3-kinase signaling
glucose transport
response to estradiol
positive regulation of multicellular organism growth
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
positive regulation of MAP kinase activity
positive regulation of insulin-like growth factor receptor signaling pathway
positive regulation of JAK-STAT cascade
positive regulation of peptidyl-tyrosine phosphorylation
growth hormone receptor signaling pathway
JAK-STAT cascade involved in growth hormone signaling pathway
bone maturation
growth hormone receptor binding
prolactin receptor binding
hormone activity
protein binding
growth factor activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000617086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000617086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000617086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | ENST00000617086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023925.hg.1 | ENST00000323322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023925.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023925.hg.1 | TC17002854.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700207231.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207231.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207231.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700207231.hg.1 | TC1700012450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700207231.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207231.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700207231.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000323322.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000342364.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000351388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000458650.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000579711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000617086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000617086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000617086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | ENST00000617086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_000515.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022559.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012450.hg.1 | NM_022560.3 | Refseq |

GH1
Genomic Map