Assay Details
| Gene Symbol: HLA-DQB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198858.1 | NP_001185787.1 | 3 - 4 | 730 | 122 | |
| GenBank mRNA | BC031995.1 | - | 3 - 4 | 667 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P19510 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P19510 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 215536_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003503.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003503.hg.1 | TC6_mcf_hap5000171.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003696.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003696.hg.1 | TC6_qbl_hap6000184.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003435.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003435.hg.1 | TC6_ssto_hap7000166.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001669.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001669.hg.1 | TC6_apd_hap1000102.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003862.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003862.hg.1 | TC6_cox_hap2000196.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002985.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002985.hg.1 | TC6_mann_hap4000164.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003697.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003697.hg.1 | TC6_qbl_hap6000184.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003434.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003434.hg.1 | TC6_ssto_hap7000166.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018717.hg.1 | ENST00000411527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018717.hg.1 | ENST00000435145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018717.hg.1 | ENST00000437316.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018717.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018717.hg.1 | TC06001571.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003863.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003863.hg.1 | TC6_cox_hap2000196.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002986.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002986.hg.1 | TC6_mann_hap4000164.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018718.hg.1 | ENST00000411527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018718.hg.1 | ENST00000435145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018718.hg.1 | ENST00000437316.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018718.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018718.hg.1 | TC06001571.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003613.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003613.hg.1 | TC6_dbb_hap3000182.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001668.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001668.hg.1 | TC6_apd_hap1000102.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003502.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003502.hg.1 | TC6_mcf_hap5000171.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003614.hg.1 | NM_001198858.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3003614.hg.1 | TC6_dbb_hap3000182.hg.1 | HTA TC |
| Human Genome U133 Plus 2.0 Array | 215536_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215536_PM_at | - | - |
| Human Genome U133A 2.0 Array | 215536_PM_at | - | - |
| Human Genome U133A 2.0 Array | 215536_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw566118 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215536_at5 | - | - |
| Human Genome U133 Set | 215536_at | - | - |
| Human Genome U133 Set | 215536_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029759 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029759 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 164735 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1741648 | AK098007.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1741648 | BC031995.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1741648 | NM_001198858.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1741648 | AK098007.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1741648 | BC031995.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1741648 | NM_001198858.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1741648 | AK098007.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1741648 | BC031995.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1741648 | NM_001198858.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1741648 | AK098007.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1741648 | BC031995.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1741648 | NM_001198858.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1741648 | AK098007.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1741648 | BC031995.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1741648 | NM_001198858.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1741648 | AK098007.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1741648 | BC031995.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1741648 | NM_001198858.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1741648 | AK098007.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1741648 | BC031995.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1741648 | NM_001198858.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1741648 | AK098007.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1741648 | BC031995.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1741648 | NM_001198858.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1741648 | AK098007.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1741648 | BC031995.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1741648 | NM_001198858.1 | human_transcriptome |

HLA-DQB2
Genomic Map