Assay Details
| Gene Symbol: ADAMTS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197941.3 | NP_922932.2 | 25 | 5440 | 150 | |
| NR_135689.1 | 26 | 5591 | 150 | |||
| XM_011543113.2 | XP_011541415.1 | 25 | 5455 | 150 | ||
| XM_011543114.2 | XP_011541416.1 | 25 | 5034 | 150 | ||
| XM_011543115.2 | XP_011541417.1 | 24 | 5343 | 150 | ||
| XM_011543116.2 | XP_011541418.1 | 23 | 5202 | 150 | ||
| XM_011543117.1 | XP_011541419.1 | 23 | 4793 | 150 | ||
| XM_011543121.2 | XP_011541423.2 | 21 | 3821 | 150 | ||
| XM_011543123.2 | XP_011541425.1 | 19 | 3559 | 150 | ||
| XM_011543124.2 | XP_011541426.1 | 16 | 3343 | 150 | ||
| XM_011543125.1 | XP_011541427.1 | 15 | 3198 | 150 | ||
| GenBank mRNA | AB209685.1 | - | 26 | 5088 | 150 | |
| AY692424.1 | - | 25 | 5398 | 150 | ||
| CR749356.1 | - | 7 | 2175 | 150 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ADAMTS6
Genomic Map