Assay Details
| Gene Symbol: GJB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198568.2 | NP_940970.1 | 2 - 3 | 319 | 72 | |
| GenBank mRNA | AK127554.1 | - | 1 - 2 | 144 | 72 | |
| BC057813.1 | - | 2 - 3 | 319 | 30336398 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001927.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK127554.1,BC057813.1,NM_198568.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06023603.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023603.hg.1 | ENST00000525899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023603.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023603.hg.1 | TC06001927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023605.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023605.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023605.hg.1 | ENST00000525899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023605.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023605.hg.1 | TC06001927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023608.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023608.hg.1 | ENST00000525899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023608.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023608.hg.1 | TC06001927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023609.hg.1 | ENST00000525899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023609.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023609.hg.1 | TC06001927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023601.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023601.hg.1 | ENST00000525899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023601.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023601.hg.1 | TC06001927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023602.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023602.hg.1 | ENST00000525899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023602.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023602.hg.1 | TC06001927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186152.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186152.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186152.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186152.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186152.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186152.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186152.hg.1 | TC0600012502.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186157.hg.1 | TC0600012502.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600186151.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186151.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186151.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186151.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186151.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186151.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186151.hg.1 | TC0600012502.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186153.hg.1 | TC0600012502.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186154.hg.1 | TC0600012502.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600186159.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186159.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186159.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186159.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186159.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186159.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186159.hg.1 | TC0600012502.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000296882.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000369576.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000442973.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012502.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186160.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186160.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186160.hg.1 | ENST00000525899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186160.hg.1 | NM_198568.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186160.hg.1 | TC0600012502.hg.1 | Clariom D TC |

GJB7
Genomic Map