Assay Details
| Gene Symbol: NRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024628.2 | NP_001019799.1 | 11 - 12 | 2389 | 75 | |
| NM_003873.5 | NP_003864.4 | 11 - 12 | 2389 | 75 | ||
| XM_006717521.1 | XP_006717584.1 | 11 - 12 | 2163 | 75 | ||
| XM_006717522.1 | XP_006717585.1 | 11 - 12 | 2163 | 75 | ||
| XM_006717523.2 | XP_006717586.1 | 11 - 12 | 2163 | 75 | ||
| XM_006717526.1 | XP_006717589.1 | 11 - 12 | 2163 | 75 | ||
| XM_017016866.1 | XP_016872355.1 | 10 - 11 | 1983 | 75 | ||
| GenBank mRNA | AB209641.1 | - | 11 - 12 | 2017 | 75 | |
| AF016050.1 | - | 11 - 12 | 1866 | 75 | ||
| AF018956.1 | - | 11 - 12 | 1866 | 75 | ||
| AF145712.1 | - | 11 - 12 | 1866 | 75 | ||
| AY249243.1 | - | 11 - 12 | 1866 | 75 | ||
| BC007533.1 | - | 11 - 12 | 2126 | 2958475 | 75 | |
| BT006995.1 | - | 11 - 12 | 1866 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001166.hg.1
Set Membership: |
|

NRP1
Genomic Map