Assay Details
| Gene Symbol: TRGC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M13231.1 | - | 5 - 6 | 791 | 111 | |
| M17323.1 | - | 429 | 111 | |||
| Gene Symbol: TARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M16804.1 | - | 5 - 6 | 769 | 111 | |
| M27331.1 | - | 2 - 3 | 382 | 111 | ||
| M30894.1 | - | 969 | 111 | |||
| Y00790.1 | - | 5 - 6 | 820 | 111 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8139101 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8139104 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8139102 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8139103 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8139100 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013539.hg.1 | ENST00000390343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013539.hg.1 | ENST00000390343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013540.hg.1 | ENST00000390348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013540.hg.1 | ENST00000390348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013540.hg.1 | ENST00000390348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010782.hg.1 | ENST00000417928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010781.hg.1 | ENST00000427089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010781.hg.1 | ENST00000427089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013540.hg.1 | ENST00000390348.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013540.hg.1 | ENST00000390348.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013540.hg.1 | ENST00000390348.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010781.hg.1 | ENST00000427089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010781.hg.1 | ENST00000427089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010782.hg.1 | ENST00000417928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013539.hg.1 | ENST00000390343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013539.hg.1 | ENST00000390343.2 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 215806_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216920_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211144_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211144_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215806_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216920_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 215806_x_at | - | - |
| Human Genome U133A 2.0 Array | 211144_x_at | - | - |
| Human Genome U133A 2.0 Array | 211144_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 215806_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 216920_s_at | - | - |
| Human Genome U133A 2.0 Array | 216920_PM_s_at | - | - |
| Human Genome U95 Set | 41468_PM_at | - | - |
| Human Genome U95 Set | 41468_at | - | - |
| Human Genome U133 Set | 215806_PM_x_at | - | - |
| Human Genome U133 Set | 216920_PM_s_at | - | - |
| Human Genome U133 Set | 211144_x_at | - | - |
| Human Genome U133 Set | 215806_x_at | - | - |
| Human Genome U133 Set | 211144_PM_x_at | - | - |
| Human Genome U133 Set | 216920_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 125726 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 141631 | - | - |

TRGC2
Genomic Map