| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005380.hg.1 |
ENST00000360359.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005380.hg.1 |
ENST00000373990.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005380.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005380.hg.1 |
TC06000510.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
ENST00000470698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007774.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600153898.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600153898.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600153898.hg.1 |
ENST00000360359.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600153898.hg.1 |
NM_015245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600153898.hg.1 |
TC0600007774.hg.1 |
Clariom D TC |