Assay Details
| Gene Symbol: GANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301409.1 | NP_001288338.1 | 7 - 8 | 1260 | 119 | |
| NM_198141.2 | NP_937784.2 | 6 - 7 | 873 | 119 | ||
| GenBank mRNA | AF545045.1 | - | 6 - 7 | 567 | 119 | |
| AF545046.1 | - | 6 - 7 | 567 | 119 | ||
| AK074037.1 | - | 6 - 7 | 765 | 119 | ||
| AK074330.1 | - | 7 - 8 | 1262 | 119 | ||
| BC059406.1 | - | 6 - 7 | 745 | 30330796 | 119 | |
| BC093833.1 | - | 6 - 7 | 590 | 7939678 | 119 | |
| BC112051.1 | - | 6 - 7 | 590 | 8327519 | 119 | |
More Information
Set Membership: |
|

GANC
Genomic Map