Assay Details
| Gene Symbol: PRSS56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195129.1 | NP_001182058.1 | 9 - 10 | 1322 | 126 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001401.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02022116.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022116.hg.1 | TC02001401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022117.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022117.hg.1 | TC02001401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179787.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179787.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179787.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179787.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179787.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179787.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179787.hg.1 | TC0200011103.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200179788.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179788.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179788.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179788.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179788.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179788.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179788.hg.1 | TC0200011103.hg.1 | Clariom D TC |

PRSS56
Genomic Map