Assay Details
| Gene Symbol: LOC105377205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058075.1 | - | 1 - 2 | 495 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bromodomain containing 1 | BRD1 |
BRL, BRPF1, BRPF2 |
-5.1kb | Sense | Chr. 22 - 49773278 - 49827927 on NCBI Build GRCh38 |
| Downstream | atherin-like | LOC105373091 | 11.8kb | Anti-sense | Chr. 22 - 49849503 - 49851083 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105377205
Genomic Map