Assay Details
| Gene Symbol: GANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301409.1 | NP_001288338.1 | 8 - 9 | 1329 | 83 | |
| NM_198141.2 | NP_937784.2 | 7 - 8 | 942 | 83 | ||
| GenBank mRNA | AF525397.1 | - | 3 - 4 | 195 | 83 | |
| AF545045.1 | - | 7 - 8 | 636 | 83 | ||
| AF545046.1 | - | 7 - 8 | 636 | 83 | ||
| AK074037.1 | - | 7 - 8 | 834 | 83 | ||
| AK074330.1 | - | 8 - 9 | 1331 | 83 | ||
| BC059406.1 | - | 7 - 8 | 814 | 30330796 | 83 | |
| BC093833.1 | - | 7 - 8 | 659 | 7939678 | 83 | |
| BC112051.1 | - | 7 - 8 | 659 | 8327519 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

GANC
Genomic Map