Assay Details
| Gene Symbol: SMC5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR936772.1 | - | 1 - 2 | 949 | 86 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500006608.hg.1 | ENST00000623883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000417795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000437091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000453876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000457998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000616965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | ENST00000621103.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012008.hg.1 | NR_110997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006608.hg.1 | ENST00000623883.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 157840 | - | - |

SMC5-AS1
Genomic Map