Assay Details
| Gene Symbol: LOC101928539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109900.1 | 2 - 3 | 3972 | 92 | ||
| GenBank mRNA | BC041836.1 | - | 2 - 3 | 3973 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928505 | LOC101928505 | -219.3kb | Anti-sense | Chr. 5 - 57650660 - 57670989 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928569 | LOC101928569 | 212.6kb | Sense | Chr. 5 - 58107632 - 58122341 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928539
Genomic Map