| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050055.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050055.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050055.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050058.hg.1 |
ENST00000368796.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050058.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050058.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050058.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050061.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050061.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050061.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050063.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050063.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050063.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050057.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050057.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050057.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050065.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050065.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050065.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050062.hg.1 |
ENST00000271820.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050062.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050062.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050062.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050059.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050059.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050059.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050064.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050064.hg.1 |
ENST00000392689.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050064.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050064.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050054.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050054.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050054.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050056.hg.1 |
ENST00000368796.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050056.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050056.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050056.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050060.hg.1 |
ENST00000271820.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050060.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050060.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050060.hg.1 |
TC01003245.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015830.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218922.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218922.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218922.hg.1 |
ENST00000368799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218922.hg.1 |
NM_002016.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218922.hg.1 |
TC0100015830.hg.1 |
Clariom D TC |