Assay Details
| Gene Symbol: OSER1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038339.2 | 1 - 2 | 121 | 137 | ||
| GenBank mRNA | BC048277.1 | - | 1 - 2 | 121 | 137 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000326.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004620.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004620.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004620.hg.1 | TC20000326.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004625.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004625.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004625.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004625.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004625.hg.1 | TC20000326.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004619.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004619.hg.1 | TC20000326.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | TC2000007447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151621.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | TC2000007447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151610.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000435163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000437730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000442383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | ENST00000623343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038338.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007447.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151608.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151608.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151608.hg.1 | NR_038339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151608.hg.1 | TC2000007447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000151608.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151608.hg.1 | ENST00000439943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151608.hg.1 | NR_038339.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 229136 | - | - |

OSER1-AS1
Genomic Map