Assay Details
| Gene Symbol: LOC101927138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC033543.1 | - | 1 - 2 | 921 | 127 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927159 | LOC101927159 | -280.2kb | Sense | Chr. 20 - 42685483 - 42688562 on NCBI Build GRCh38 | |
| Downstream | serine and arginine rich splicing factor 6 | SRSF6 |
B52, HEL-S-91, SFRS6, SRP55 |
486.4kb | Sense | Chr. 20 - 43457864 - 43463605 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927138
Genomic Map