Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 30 member 9 | SLC30A9 |
C4orf1, GAC63, HUEL, ZNT9 |
-8.3kb | Sense | Chr. 4 - 41990472 - 42087534 on NCBI Build GRCh38 |
| Downstream | BEN domain containing 4 | BEND4 | CCDC4 | 11.5kb | Anti-sense | Chr. 4 - 42110853 - 42152878 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map