Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoprotein membrane anchor with glycosphingolipid microdomains 1 | PAG1 |
CBP, PAG |
-172.6kb | Sense | Chr. 8 - 80967810 - 81112080 on NCBI Build GRCh38 |
| Downstream | peripheral myelin protein 2 | PMP2 |
FABP8, M-FABP, MP2, P2 |
155.2kb | Sense | Chr. 8 - 81440326 - 81447484 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map