| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030395.hg.1 |
ENST00000360064.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030395.hg.1 |
ENST00000366574.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030395.hg.1 |
ENST00000540213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030395.hg.1 |
ENST00000542288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030395.hg.1 |
ENST00000542537.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030395.hg.1 |
NM_001035.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030395.hg.1 |
TC01001957.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
ENST00000360064.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
ENST00000366574.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
ENST00000609119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
NM_001035.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
ENST00000360064.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
ENST00000366574.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
ENST00000609119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
NM_001035.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189287.hg.1 |
TC0100012113.hg.1 |
Clariom D TC |