Assay Details
| Gene Symbol: SOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005633.3 | NP_005624.2 | 20 - 21 | 3383 | 151 | |
| XM_011533062.2 | XP_011531364.1 | 20 - 21 | 3551 | 151 | ||
| XM_011533064.2 | XP_011531366.1 | 20 - 21 | 3291 | 151 | ||
| XM_011533066.2 | XP_011531368.1 | 13 - 14 | 2292 | 151 | ||
| GenBank mRNA | AK290228.1 | - | 21 - 22 | 3429 | 151 | |
| L13857.1 | - | 20 - 21 | 3342 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001761.hg.1
Set Membership: |
|

SOS1
Genomic Map