Assay Details
| Gene Symbol: DNASE1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004944.3 | NP_004935.1 | 3 - 4 | 415 | 156 | |
| GenBank mRNA | AF047354.1 | - | 3 - 4 | 388 | 156 | |
| AK131323.1 | - | 6 - 7 | 910 | 156 | ||
| AK313303.1 | - | 3 - 4 | 381 | 156 | ||
| BC015831.1 | - | 3 - 4 | 385 | 4723725 | 156 | |
| U56814.1 | - | 3 - 4 | 342 | 156 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001498.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027307.hg.1 | ENST00000318316.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027307.hg.1 | ENST00000450710.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027307.hg.1 | ENST00000483681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027307.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027307.hg.1 | TC03001498.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184746.hg.1 | TC0300011298.hg.1 | Clariom D TC |

DNASE1L3
Genomic Map