Assay Details
| Gene Symbol: DNASE1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI834035.1 | - | 1 - 2 | 374 | 125 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |

DNASE1L3
Genomic Map