Assay Details
| Gene Symbol: DNASE1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131323.1 | - | 3 - 4 | 497 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131323.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000394549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000460422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000461914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000463694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000477209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000483681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | ENST00000486455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_001256560.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011298.hg.1 | NM_004944.3 | Refseq |

DNASE1L3
Genomic Map