Assay Details
| Gene Symbol: FCRLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184870.1 | NP_001171799.1 | 1 - 2 | 370 | 82 | |
| GenBank mRNA | AF329494.1 | - | 1 - 2 | 77 | 82 | |
More Information
Set Membership: |
|

FCRLA
Genomic Map