Assay Details
| Gene Symbol: FCRLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184873.1 | NP_001171802.1 | 2 - 3 | 526 | 79 | |
| GenBank mRNA | AF390036.1 | - | 2 - 3 | 203 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001411.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000236938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000350710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000367949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000367950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000367953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000367957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000367959.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000470841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000540521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000540926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | ENST00000546024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | NM_001184866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | NM_001184867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | NM_001184870.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | NM_001184871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | NM_001184872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | NM_001184873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | NM_032738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022080.hg.1 | TC01001411.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000236938.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000350710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000367949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000367953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000367957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000367959.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000470841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000540521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000540926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | ENST00000546024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | NM_001184866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | NM_001184867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | NM_001184870.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | NM_001184871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | NM_001184872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | NM_001184873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | NM_032738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022081.hg.1 | TC01001411.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000236938.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000350710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000367949.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000367953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000367957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000367959.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000470841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000540521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000546024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184867.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184870.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184873.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_032738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000236938.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000350710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000367949.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000367953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000367957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000367959.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000470841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000540521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | ENST00000546024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184867.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184870.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_001184873.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | NM_032738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176331.hg.1 | TC0100010365.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000236938.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000294796.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000309691.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000349527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000350710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367949.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367959.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000470841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000540521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000546024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184867.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184870.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184873.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_032738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000236938.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000294796.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000309691.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000349527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000350710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367949.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000367959.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000470841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000540521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | ENST00000546024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184867.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184870.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_001184873.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | NM_032738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176328.hg.1 | TC0100010365.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000236938.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000350710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367949.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367959.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000470841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000540521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000546024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184867.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184870.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184873.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_032738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000236938.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000350710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367949.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000367959.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000470841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000540521.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | ENST00000546024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184867.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184870.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_001184873.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | NM_032738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176330.hg.1 | TC0100010365.hg.1 | Clariom D TC |

FCRLA
Genomic Map