| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000236938.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000309691.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000349527.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000367949.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000367950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000367953.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000367959.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000470841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000540926.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
ENST00000546024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
NM_001184866.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
NM_001184867.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
NM_001184873.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
NM_032738.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022073.hg.1 |
TC01001411.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000236938.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000294796.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000309691.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000367953.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000367957.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000367959.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000470841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000540521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000540926.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
ENST00000546024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
NM_001184866.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
NM_001184867.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
NM_001184870.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
NM_001184872.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
NM_032738.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01022077.hg.1 |
TC01001411.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000236938.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000294796.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000309691.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000367953.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000367957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000367959.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000470841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000540521.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000546024.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_001184866.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_001184867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_001184870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_001184872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_032738.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000236938.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000294796.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000309691.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000367953.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000367957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000367959.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000470841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000540521.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
ENST00000546024.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_001184872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_001184866.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_001184867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_001184870.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
NM_032738.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176326.hg.1 |
TC0100010365.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000236938.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000309691.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000349527.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000367949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000367950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000367953.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000367959.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000470841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000546024.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
NM_001184866.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
NM_001184867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
NM_001184873.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
NM_032738.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000236938.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000309691.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000349527.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000367949.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000367950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000367953.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000367959.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000470841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
ENST00000546024.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
NM_001184866.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
NM_001184867.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
NM_001184873.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
NM_032738.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176322.hg.1 |
TC0100010365.hg.1 |
Clariom D TC |