Assay Details
| Gene Symbol: SNU13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028811.1 | XP_016884300.1 | 2 - 3 | 247 | 85 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000215956.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000215956.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000215956.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000215956.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000215956.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000215956.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000215956.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000215956.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000355257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000355257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000355257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000355257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000355257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000355257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000355257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000401959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000402458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000402458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000402458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000463675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000463675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000463675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000463675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000469028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000469028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000469028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000469522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000469522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000469522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000488571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000488571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | ENST00000488571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_001003796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_001003796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_001003796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_001003796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_001003796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_001003796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_001003796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008837.hg.1 | NM_005008.3 | Refseq |

SNU13
Genomic Map