| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009654.hg.1 |
ENST00000372070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009654.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009654.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009654.hg.1 |
TC01000574.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009657.hg.1 |
ENST00000351829.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009657.hg.1 |
ENST00000372070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009657.hg.1 |
ENST00000471651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009657.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009657.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009657.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009657.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009657.hg.1 |
TC01000574.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009655.hg.1 |
ENST00000372070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009655.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009655.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009655.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009655.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009655.hg.1 |
TC01000574.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009651.hg.1 |
ENST00000372070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009651.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009651.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009651.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009651.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01009651.hg.1 |
TC01000574.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
ENST00000372070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
ENST00000434299.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
ENST00000621846.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
ENST00000372070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
ENST00000434299.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
ENST00000621846.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158011.hg.1 |
TC0100008126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
ENST00000372070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
ENST00000476957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
ENST00000496999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
ENST00000372070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
ENST00000476957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
ENST00000496999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158014.hg.1 |
TC0100008126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000372070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000434299.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000476957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000496999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000621846.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000372070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000434299.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000476957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000496999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
ENST00000621846.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158015.hg.1 |
TC0100008126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000351829.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000372070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000434299.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000471651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000476957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000481885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000496999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000621846.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000351829.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000372070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000434299.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000471651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000476957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000481885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000496999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
ENST00000621846.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
NM_001202413.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
NM_001202414.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
NM_006066.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
NM_153326.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100158017.hg.1 |
TC0100008126.hg.1 |
Clariom D TC |