Assay Details
| Gene Symbol: COL9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001852.3 | NP_001843.1 | 12 - 13 | 699 | 68 | |
| XM_006710365.3 | XP_006710428.1 | 13 - 14 | 807 | 68 | ||
| XM_011540715.2 | XP_011539017.1 | 12 - 13 | 1287 | 68 | ||
| XM_011540716.2 | XP_011539018.1 | 11 - 12 | 767 | 68 | ||
| XM_011540717.2 | XP_011539019.1 | 11 - 12 | 1615 | 68 | ||
| XM_017000332.1 | XP_016855821.1 | 12 - 13 | 753 | 68 | ||
| XM_017000333.1 | XP_016855822.1 | 11 - 12 | 753 | 68 | ||
| GenBank mRNA | BC041479.1 | - | 11 - 12 | 932 | 68 | |
| BC136326.1 | - | 12 - 13 | 692 | 9020313 | 68 | |
| BC136327.1 | - | 12 - 13 | 692 | 9020314 | 68 | |
| M95610.1 | - | 12 - 13 | 588 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002554.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |

COL9A2
Genomic Map