Assay Details
| Gene Symbol: COL9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540716.2 | XP_011539018.1 | 1 - 2 | 271 | 93 | |
| XM_017000333.1 | XP_016855822.1 | 1 - 2 | 269 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011540716.2, XM_017000333.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372736.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000372748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000417105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000427563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000461118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000466267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000482722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000488463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000490132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000495948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | ENST00000496215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013823.hg.1 | NM_001852.3 | Refseq |

COL9A2
Genomic Map