Assay Details
| Gene Symbol: FH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544132.2 | XP_011542434.1 | 1 - 2 | 635 | 143 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011544132.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000493477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000493477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000493477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000497042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000497042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | ENST00000497042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000366560.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000493477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000493477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000493477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000497042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000497042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | ENST00000497042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017945.hg.1 | NM_000143.3 | Refseq |

FH
Genomic Map